missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-methylpyridine, 98%

Product Code. 15462679
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462679 5 g 5g
15472679 250 mg 250mg
15452679 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462679 Supplier Thermo Scientific Alfa Aesar Supplier No. H65997.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6940-57-4 | C8H9NO | 135.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-57-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD06658293
InChI Key FPQMUQPPAYCAME-UHFFFAOYSA-N
Synonym 2-acetyl-6-methylpyridine, 1-6-methylpyridin-2-yl ethanone, 1-6-methylpyridin-2-yl ethan-1-one, ketone, methyl 6-methyl-2-pyridyl, 6-acetyl-2-picoline, 1-6-methyl-pyridin-2-yl-ethanone, 1-6-methyl-2-pyridinyl ethanone, 6-methyl-2-acetylpyridine, ethanone, 1-6-methyl-2-pyridinyl, 6-acetyl-2-methylpyridine
PubChem CID 138872
IUPAC Name 1-(6-methylpyridin-2-yl)ethanone
SMILES CC1=CC=CC(=N1)C(=O)C

Specifications

Color Yellow
Boiling Point 209°C
Flash Point 84°C (183°F)
Refractive Index 1.515
Quantity 5 g
Beilstein 109564
Formula Weight 135.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Acetyl-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.