missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylthiophene, 98%

Product Code. 11463644
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463644 5 g 5g
11473644 25 g 25g
11483644 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463644 Supplier Thermo Scientific Alfa Aesar Supplier No. A11282.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13679-74-8 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-74-8
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00014529
InChI Key YOSDTJYMDAEEAZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
PubChem CID 83655
IUPAC Name 1-(5-methylthiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(C)S1

Specifications

Melting Point 24°C to 28°C
Density 1.119
Boiling Point 65°C to 67°C (1 mmHg)
Refractive Index 1.5625
Quantity 5 g
Beilstein 110854
Sensitivity Light sensitive
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Acetyl-5-methylthiophene
compliance-icons
Product Identifier
  • 2-Acetyl-5-methylthiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 1-(5-Methyl-2-thienyl)ethan-1-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.