missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylfuran, 98+%

Product Code. 11423237
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423237 10 g 10g
11494040 25 g 25g
11433237 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423237 Supplier Thermo Scientific Alfa Aesar Supplier No. A16172.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1193-79-9 | C7H8O2 | 124.139 g/mol

Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-79-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00003243
InChI Key KEFJLCGVTHRGAH-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone
PubChem CID 14514
ChEBI CHEBI:562752
IUPAC Name 1-(5-methylfuran-2-yl)ethanone
SMILES CC1=CC=C(O1)C(=O)C

Specifications

Density 1.068
Boiling Point 100°C to 101°C (25 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.513
Quantity 10 g
Beilstein 110853
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 2-Acetyl-5-methylfuran
compliance-icons
Product Identifier
  • 2-Acetyl-5-methylfuran
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2-Acetyl-5-methylfuran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.