missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetyl-5-iodo-2H-indazole, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 1174064-60-8 |
Assay Percent Range | 95% |
Molecular Formula | C9H7IN2O |
MDL Number | MFCD11520128 |
Quantity | 1 g |
Synonym | 2-acetyl-5-iodo-2h-indazole, 1-5-iodoindazol-2-yl ethanone, 1-5-iodo-2h-indazol-2-yl ethan-1-one, 1-5-iodo-2h-indazol-2-yl ethanone |
InChI Key | JXZRYFOLDPVLCP-UHFFFAOYSA-N |
SMILES | CC(=O)N1C=C2C=C(I)C=CC2=N1 |
IUPAC Name | 1-(5-iodoindazol-2-yl)ethanone |
Molecular Weight (g/mol) | 286.07 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'