missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-bromothiophene, 99%

Product Code. 10083610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083610 25 g 25g
10567763 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083610 Supplier Thermo Scientific Acros Supplier No. 308330250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5370-25-2 | C6H5BrOS | 205.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5370-25-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD00014528
InChI Key IGBZCOWXSCWSHO-UHFFFAOYSA-N
Synonym 2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
PubChem CID 79335
IUPAC Name 1-(5-bromothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Yellow
Boiling Point 103°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 205.07
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2-Acetyl-5-bromothiophene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.