missing translation for 'onlineSavingsMsg'
Learn More

(2-Acetyl-4-methylpentyl)trimethylammonium Iodide, TRC

Product Code. 30315747
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315747 Supplier Toronto Research Chemicals Supplier No. TRCA1645501G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Acetyl-4-methylpentyl)trimethylammonium Iodide
CAS 1069-62-1
Molecular Formula C11H24INO
Quantity 1 g
Synonym 2-Acetyl-N,N,N,4-tetramethyl-1-pentanaminium Iodide
InChI Formula InChI=1S/C11H24NO.HI/c1-9(2)7-11(10(3)13)8-12(4,5)6;/h9,11H,7-8H2,1-6H3;1H/q+1;/p-1
SMILES O=C(C(C[N+](C)(C)C)CC(C)C)C.[I-]
IUPAC Name (2-acetyl-4-methylpentyl)-trimethylazanium;iodide
Molecular Weight (g/mol) 313.22
Formula Weight 313.0903
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.