missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-methylpyrazine, 98+%, ACROS Organics™

Product Code. 11330786 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330786 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11330786 Supplier Thermo Scientific Acros Supplier No. 264850025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 23787-80-6
CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Molecular Formula C7H8N2O
Refractive Index 1.5206 to 1.5226 (20°C, 589nm)
MDL Number MFCD00014612
Quantity 2.5 g
Synonym 2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
InChI Key QUNOTZOHYZZWKQ-UHFFFAOYSA-N
SMILES CC(=O)C1=NC=CN=C1C
IUPAC Name 1-(3-methylpyrazin-2-yl)ethan-1-one
Molecular Weight (g/mol) 136.15
PubChem CID 32093
Formula Weight 136.15
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 2-Acetyl-3-methylpyrazine, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.