missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-naphthol, 99%

Product Code. 10403115 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10403115

Brand: Thermo Scientific Acros 195000050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 711-79-5 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 711-79-5
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003963
InChI Key JBGJVMVWYWUVOW-UHFFFAOYSA-N
Synonym 1'-hydroxy-2'-acetonaphthone, 2-acetyl-1-naphthol, 1-hydroxy-2-acetylnaphthalene, 2-acetyl-1-hydroxynaphthalene, 1-hydroxy-2-acetonaphthone, ethanone, 1-1-hydroxy-2-naphthalenyl, 1-1-hydroxynaphthalen-2-yl ethanone, 1-1-hydroxy-2-naphthalenyl ethanone, 2'-acetonaphthone, 1'-hydroxy, 1-hydroxy-2-naphthyl methyl ketone
PubChem CID 69733
IUPAC Name 1-(1-hydroxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC=C2C=CC=CC2=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 97°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC10H6COCH3
Quantity 5 g
Beilstein 08, 149
Formula Weight 186.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-1-naphthol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetyl-1-naphthol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.