missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidophenol, TRC

Product Code. 17972535
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972535 10 g 10g
17954675 1 g 1g
17920585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972535 Supplier Toronto Research Chemicals Supplier No. TRCA15849010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-80-2
Molecular Formula C8 H9 N O2
Molecular Weight (g/mol) 151.16
Synonym N-(2-Hydroxyphenyl)-acetamide, Paracetamol Imp. A (EP), 2-Acetamidophenol, Acetamide, N-(2-hydroxyphenyl)-, Acetanilide, 2'-hydroxy- (7CI,8CI), Acetanilide, o-hydroxy- (3CI), 2-(Acetylamino)phenol, 2-(N-Acetylamino)phenol, 2-Acetaminophenol, 2-Hydroxyacetanilide, 2'-Hydroxyacetanilide, N-Acetyl-2-aminophenol, N-Acetyl-o-aminophenol, NSC 3989, Orthocetamol, o-(Acetylamino)phenol, o-Acetamidophenol, o-Acetaminophenol, o-Hydroxyacetanilide
IUPAC Name N-(2-hydroxyphenyl)acetamide
SMILES CC(=O)Nc1ccccc1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H9NO2/c1-6(10)9-7-4-2-3-5-8(7)11/h2-5,11H,1H3,(H,9,10)
Formula Weight 151.0633
Chemical Name or Material N-(2-Hydroxyphenyl)-acetamide (2-Acetamidophenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.