missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%

Product Code. 10132324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132324 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132324 Supplier Thermo Scientific Acros Supplier No. 148360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 69812-29-9 | C6H7ClN2O3S2 | 254.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 69812-29-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 161°C to 164°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H7ClN2O3S2
MDL Number MFCD00005320
Quantity 1 g
Synonym 2-acetamido-4-methylthiazole-5-sulfonyl chloride, 2-acetamido-4-methyl-5-thiazolesulfonyl chloride, 2-acetylamino-4-methyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 2-acetylamino-4-methyl, 2-acetamido-4-methylthiazole-5-sulphonyl chloride, 2-acetylamino-4-methyl-1,3-thiazole-5-sulfonyl chloride, 2-acetylamino-4-methylthiazole-5-sulfonyl chloride, 2-acetylamino-4-methyl-1,3-thiazol-5-ylsulphonyl chloride, n-5-chlorosulfonyl-4-methyl-1,3-thiazol-2-yl acetamide, ksc355a5n
InChI Key NXGKPRKPUCSEIL-UHFFFAOYSA-N
SMILES CC1=C(SC(=N1)NC(=O)C)S(=O)(=O)Cl
IUPAC Name 2-acetamido-4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol) 254.71
PubChem CID 96951
Formula Weight 254.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-4-methyl-5-thiazolesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetamido-4-methyl-5-thiazolesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.