missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Hydroxy-3-oxo-4-oxanonyl)-2-methyl-1,2,3,4-tetrahydropapaverinium Benzenesulfonate, TRC

Product Code. 30395153
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395153 50 mg -
30389201 100 mg -
30403353 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395153 Supplier Toronto Research Chemicals Supplier No. TRCH80533550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C29H42NO7+
Molecular Weight (g/mol) 516.6
Synonym 1-[(3,4-Dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-2-[3-[(5-hydroxypentyl)oxy]-3-oxopropyl]-6,7-dimethoxy-2-methylisoquinolinium Benzenesulfonate, Atracurium Impurity D
SMILES COc1ccc(CC2c3cc(OC)c(OC)cc3CC[N+]2(C)CCC(=O)OCCCCCO)cc1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H42NO7/c1-30(14-12-29(32)37-16-8-6-7-15-31)13-11-22-19-27(35-4)28(36-5)20-23(22)24(30)17-21-9-10-25(33-2)26(18-21)34-3/h9-10,18-20,24,31H,6-8,11-17H2,1-5H3/q+1
Chemical Name or Material 2-(9-Hydroxy-3-oxo-4-oxanonyl)-2-methyl-1,2,3,4-tetrahydropapaverinium Benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.