missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-5,12-dihydroquinolino[2,3-b]acridine-7,14-dione (Technical Grade), TRC

Product Code. 30369908
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369908 100 mg -
30384788 500 mg 500mg
30372669 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369908 Supplier Toronto Research Chemicals Supplier No. TRCD481185100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 980-26-7
Molecular Formula C22H16N2O2
Molecular Weight (g/mol) 340.37
Synonym 5,12-Dihydro-2,9-dimethylquino[2,3-b]acridine-7,14-dione, 2,9-Dimethyl-5,12-dihydroquinolino[2,3-b]acridine-7,14-dione, 2,9-Dimethylquinacridone, 2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione, Acramin Scarlet LDCN, Aquatone Red 122, Bayscript Magenta VP-SP 25012, C.I. 73915, C.I. PR 122, C.I. Pigment Red 122, CFR 002, CFR 130P, CFR 321, CFR 338-3, COJ 465M, CR 759D, Cab-O-Jet 260, Cab-O-Jet 266, Cab-O-Jet 465M, Cab-O-Jet IJX 266, Cab-O-Jet IJX 462M, Cabot 465M, Cabot IJX 462M, Chromofine Magenta 6886, Chromofine Magenta 6887, Chromofine Magenta RG, Chromofine Red 321, Chromofine Red 6111, Chromofine Red 6111T, Chromofine Red 6112JC, Chromophthal Jet Magenta DMQ, Cinquasia K 4410, Cinquasia Magenta D 4550J, Cinquasia Pink K 4410, Cinquasia Red RT 355, Clariant R 122EO2VP2621, Colortech Red RD 1221, Colortex Red UG 276, Colortex Red UG 515, Cromophtal Jet Magenta DMQ, Cromophtal Jet Magenta DMQ-PR 122, Cromophtal Jet Magenta DWQ, Cromophtal Pink PT, Dimethylquinacridone, ECR 184, ECR 185, ECR 187, ECR 187 (pigment), EP 1000, EP 1000 (pigment), Emacol SF Blue H 502F, Eupolen PE Red 47-9001, Eupolen Red 47-9001, Fastogen Magenta R, Fastogen Magenta RTS, Fastogen Magenta RY, Fastogen Super Magenta R, Fastogen Super Magenta RE 01, Fastogen Super Magenta RE 03, Fastogen Super Magenta RE 05, Fastogen Super Magenta RG, Fastogen Super Magenta RGT, Fastogen Super Magenta RH, Fastogen Super Magenta RS, Fastogen Super Magenta RTS, Fastogen Super Magenta RTSP, Fastogen Super Magenta RY, HPC Red 1220, HiFast N Conc Fuchsia, High Performance Colors PR 1220, Hostacopy E 02M101, Hostajet EPT Magenta, Hostajet Magenta E-PT, Hostapem Rosa E, Hostaperm E 01, Hostaperm Pink E, Hostaperm Pink E 02, Hostaperm Pink E Transparent 01, Hostaperm Pink E-WD, Hostaperm Pink EB, Hostaperm Pink EB Transparent, Hostaperm Red E-WD, Hostaperm Rose E, Hostatint Pink A-EB, Hostatint Pink E 30, IJX 266, IJX 462M, Ink Jet Magenta E 02, Ink Jet Magenta E 02VP261, Ink Jet Magenta E 02VP2621, Ink Jet Magenta E-S, Inxel Red A 303, Irgaphor Magenta DMQ, Jet Magenta DMQ, KET Red 309, KET Red 310, KF Red 1, Keystone Jet Print Micro Magenta, Liogen Magenta RR 122, Lionogen Magenta 5750, Lionogen Magenta 5790, Lionogen Magenta 5793, Lionogen Magenta FG 5793, Lionogen Magenta ID 120, Lionogen Magenta R, Magenta E 02VP2621, Magenta QWD 0108, Magenta RG, Magenta RT 150DL, Monolite Rubine 3B, PR 122, PR 122RGT, PR 124, PV Fast Pink E, PV Fast Pink EB, PV Fast Pink EB trans, Paliogen Red 4790, Paliogen Red L 4790, Permanent Pink E, Pigmatex Fuchsia BW, Pigment Magenta 122, Pigment Red 122, Pigment Red 1221D, Pink E, Pink E 02, Pro-Jet APD 4000 Magenta, QFD 1146, QJD 3122, QWD 0108 Magenta, Quinacridone Magenta, Quinacridone Red 122A, Quinacridone Red F, Quindo Magenta RV 6704, Quindo Magenta RV 6803, Quindo Magenta RV 6828, Quindo Magenta RV 6831, Quindo Magenta RV 6832, RTS, RV 6828, Sanyo Pigment Red 3090, Sensijet Ultra Magenta PR 122, Sun Quinacridone Magenta 122PE, Sunfast Magenta, Sunfast Magenta 122, Sunfast Magenta 228-0013, Sunfast Magenta Presscake 122, Super Magenta R, Super Magenta RG, Super Magenta RY, Symuler Super Magenta RGT, Symuler Super Magenta RY, TCR 12203IJ, TRM 11, Toner Magenta E, Toner Magenta E 002, Toner Magenta E 02, Toner Magenta EB, Toner Magenta EO 02, Vynamon Red 312201, W 92930, Xfast Magenta 4790
IUPAC Name 2,9-dimethyl-5,12-dihydroquinolino[2,3-b]acridine-7,14-dione
SMILES Cc1ccc2Nc3cc4C(=O)c5cc(C)ccc5Nc4cc3C(=O)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H16N2O2/c1-11-3-5-17-13(7-11)21(25)15-9-20-16(10-19(15)23-17)22(26)14-8-12(2)4-6-18(14)24-20/h3-10H,1-2H3,(H,23,25)(H,24,26)
Formula Weight 340.1212
Chemical Name or Material 2,9-Dimethyl-5,12-dihydroquinolino[2,3-b]acridine-7,14-dione (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.