missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%

Product Code. 11410233
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410233 5 g 5g
11420233 25 g 25g
11404050 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11398.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol

2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.

Solubility
It is soluble in methanol: 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34302-69-7
Molecular Formula C28H26N4O
Molecular Weight (g/mol) 434.54
MDL Number MFCD00149306,MFCD00004973,MFCD00149306,MFCD23140843
InChI Key IEBXFSLFDFHSRD-UHFFFAOYSA-N
Synonym unii-2sdt9ev86w, 2sdt9ev86w, neocuproine hemihydrate mi, 1,10-phenanthroline, 2,9-dimethyl-, hemihydrate, 1,10-phenanthroline, 2,9-dimethyl-, hydrate 2:1, 2,9-dimethyl-1,10-phenthroline hemihydrate
PubChem CID 67652146
IUPAC Name 2,9-dimethyl-1,10-phenanthroline;hydrate
SMILES O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

Melting Point 160°C to 164°C
Quantity 5 g
Beilstein 153179
Merck Index 14,6449
Solubility Information It is soluble in methanol: 0.1g/mL.
Formula Weight 217.27 (208.26 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.