missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Carbazolyl)ethylboronic acid pinacol ester, 98%

Product Code. p-7047661
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11335355

Brand: Thermo Scientific Alfa Aesar L19580.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608534-41-4 | C20H24BNO2 | 321.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608534-41-4
Molecular Formula C20H24BNO2
Molecular Weight (g/mol) 321.227
MDL Number MFCD03788738
InChI Key NTUZDLWSQJRXAM-UHFFFAOYSA-N
Synonym 2-9-carbazolyl ethylboronic acid pinacol ester, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl-9h-carbazole, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 9-2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 2-2-9h-carbazole-9-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 45925651
IUPAC Name 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)CCN2C3=CC=CC=C3C4=CC=CC=C42

Specifications

Melting Point 111°C to 113°C
Quantity 250 mg
Formula Weight 321.23
Percent Purity 98%
Chemical Name or Material 2-(9-Carbazolyl)ethylboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.