missing translation for 'onlineSavingsMsg'
Learn More

2-(8-Chloro-1-naphthylthio)acetic acid, 98%, Thermo Scientific™

Product Code. 11484733
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484733 5 g 5g
11474733 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484733 Supplier Thermo Scientific Alfa Aesar Supplier No. A11634.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 129-94-2
Molecular Formula C12H8ClO2S
Molecular Weight (g/mol) 251.70
MDL Number MFCD00043901
InChI Key WPNAGQJVWAFQJV-UHFFFAOYSA-M
Synonym 8-chloro-1-naphthylthio acetic acid, 2-8-chloro-1-naphthylthio acetic acid, 2-8-chloronaphthalen-1-yl sulfanyl acetic acid, 2-8-chloronaphthalen-1-yl thio acetic acid, 8-chloronaphthalen-1-yl sulfanyl acetic acid, 2-8-chloro-1-naphthyl thio acetic acid, maybridge1_001748, 8-chloro-1-naphthyl sulfanyl acetic acid, acmc-1bugo, 2-8-chloronaphthylthio acetic acid
PubChem CID 67219
IUPAC Name 2-(8-chloronaphthalen-1-yl)sulfanylacetic acid
SMILES [O-]C(=O)CSC1=C2C(Cl)=CC=CC2=CC=C1

Specifica

Melting Point 155°C to 157°C
Quantity 5 g
Formula Weight 252.72
Percent Purity 98%
Chemical Name or Material 2- (8-Chloro-1-naphthylthio)acetic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.