missing translation for 'onlineSavingsMsg'
Learn More

2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%

Product Code. 11364444
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364444 5 g 5g
11354444 1 g 1g
11374444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364444 Supplier Thermo Scientific Alfa Aesar Supplier No. L01081.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50816-20-1 | C13H25BrO2 | 293.245 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50816-20-1
Molecular Formula C13H25BrO2
Molecular Weight (g/mol) 293.245
MDL Number MFCD00014648
InChI Key JCRBYQZIJFWGOO-UHFFFAOYSA-N
Synonym 2-8-bromooctyl oxy tetrahydro-2h-pyran, 2h-pyran, 2-8-bromooctyl oxy tetrahydro, 1-bromo-8-tetrahydropyranyloxy octane, 8-bromooctyl tetrahydropyranyl ether, 1-bromo-8-tetrahydropyranyloxyoctane, 2-8-bromooctyloxy tetrahydropyran, 2-8-bromooctyl oxy oxane, acmc-20aowo
PubChem CID 170916
IUPAC Name 2-(8-bromooctoxy)oxane
SMILES C1CCOC(C1)OCCCCCCCCBr

Specifications

Boiling Point 130°C to 132°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.477
Quantity 5 g
Beilstein 1306644
Formula Weight 293.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-(8-Bromooctyloxy)tetrahydropyran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.