missing translation for 'onlineSavingsMsg'
Learn More

2,8-Bis(trifluoromethyl)-4-quinolinol, 99%, Thermo Scientific™

Catalog Number 10207230
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
35853-41-9
C11H5F6NO
281.15
MFCD00075091
JIWHKBAFGFPZKM-UHFFFAOYSA-N
2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
736140
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
This item is not returnable. View return policy
35853-41-9
C11H5F6NO
281.15
MFCD00075091
JIWHKBAFGFPZKM-UHFFFAOYSA-N
2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
736140
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
CAS Min % 98.5
CAS Max % 100.0
Melting Point 130.0°C to 134.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 281.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 8-Bis(trifluoromethyl)-4-quinolinol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'