missing translation for 'onlineSavingsMsg'
Learn More

2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, 97%

Product Code. 10455769 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10455769

Brand: Thermo Scientific Acros 428250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 331465-71-5 | C18H39N4P | 342.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331465-71-5
Molecular Formula C18H39N4P
Molecular Weight (g/mol) 342.51
MDL Number MFCD03840347
InChI Key WFHPXSHLCFHEIA-UHFFFAOYSA-N
Synonym 2,8,9-triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, triisobutylphosphatrane, 2,8,9-tris 2-methylpropyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, acmc-1bmup, 4,6,11-tris 2-methylpropyl-1,4,6,11-tetraza-5-phosphabicyclo 3.3.3 undecane, nitrilotrisethylenetris isobutylimino phosphine, 2,8,9-tri-i-butyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, 2,8,9-triisobutyl-2,5,8,9-tetraaza-1-phospha-bicyclo 3.3.3 undecane
PubChem CID 4571243
IUPAC Name 4,6,11-tris(2-methylpropyl)-1,4,6,11-tetraza-5-phosphabicyclo[3.3.3]undecane
SMILES CC(C)CN1CCN2CCN(P1N(CC2)CC(C)C)CC(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9640g/mL
Elemental Analysis 60.60 to 65.65% C, 11.02 to 11.94% H, 15.70 to 17.01% N
Boiling Point 110.0°C to 118.0°C (0.5 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10155
Specific Gravity 0.964
Solubility Information Solubility in water: insoluble
Formula Weight 342.51
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Show More Show Less
compliance-icons
Product Identifier
  • 2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo¢3.3.3!undecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.