missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethylnaphthalene, 99%

Product Code. 11480297
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480297 0.25 g 0.25g
11470297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480297 Supplier Thermo Scientific Alfa Aesar Supplier No. B21704.01
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 582-16-1 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-16-1
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004121
InChI Key LRQYSMQNJLZKPS-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dimethyl, 2,7-dmn, unii-on63xh5bq4, 2,7-dimethyl-naphthalene, on63xh5bq4, acmc-1ar8i, 2,7-dimethylnaphthalene, naphthalene, 2,7-dimethyl-8ci 9ci
PubChem CID 11396
ChEBI CHEBI:48632
IUPAC Name 2,7-dimethylnaphthalene
SMILES CC1=CC2=CC(C)=CC=C2C=C1

Specifications

Melting Point 96°C to 98°C
Boiling Point 262°C to 264°C
Quantity 0.25 g
Beilstein 1852737
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material 2,7-Dimethylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.