missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethyl-10,11-dihydropyrimido[2',1':2,3]imidazo[4,5-f]quinoxalin-9(7H)-one, TRC

Product Code. 30387471
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387471 5 mg 5mg
30373379 10 mg 10mg
30371675 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387471 Supplier Toronto Research Chemicals Supplier No. TRCD4622085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,7-Dimethyl-10,11-dihydropyrimido[2',1':2,3]imidazo[4,5-f]quinoxalin-9(7H)-one
Analyte or Component Names 2,7-Dimethyl-10,11-dihydropyrimido[2',1':2,3]imidazo[4,5-f]quinoxalin-9(7H)-one
Shipping Condition Room Temperature
Molecular Formula C14 H13 N5 O
InChI Formula InChI=1S/C14H13N5O/c1-8-7-15-9-3-4-10-13(12(9)16-8)19-6-5-11(20)17-14(19)18(10)2/h3-4,7H,5-6H2,1-2H3
Solution Type Neat
SMILES CN1C2=NC(=O)CCN2c3c1ccc4ncc(C)nc34
IUPAC Name 4,11-dimethyl-3,6,11,13,17-pentazatetracyclo[8.7.0.02,7.012,17]heptadeca-1(10),2,4,6,8,12-hexaen-14-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 267.286
Quantity 5 mg
Formula Weight 267.112
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.