missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dihydroxynaphthalene, 97%

Product Code. 11433923
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433923 50 g 50g
11443923 250 g 250g
11414050 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433923 Supplier Thermo Scientific Alfa Aesar Supplier No. A10229.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 582-17-2 | C10H8O2 | 160.17 g/mol

2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-17-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004085
InChI Key DFQICHCWIIJABH-UHFFFAOYSA-N
Synonym 2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene
PubChem CID 11397
IUPAC Name naphthalene-2,7-diol
SMILES OC1=CC2=CC(O)=CC=C2C=C1

Specifications

Melting Point 183°C to 188°C
Quantity 50 g
Beilstein 2042383
Solubility Information Soluble in water.
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 2,7-Dihydroxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.