missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2',7'-Dichlorofluorescein, pure

Product Code. 10032600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032600 5 g Glass bottle 5g
10511451 25 g Glass bottle 25g
10105113 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10032600 Supplier Thermo Scientific Chemicals Supplier No. 191530050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-54-0
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.195
MDL Number MFCD00005047
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
Synonym 2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
PubChem CID 64944
ChEBI CHEBI:51596
IUPAC Name 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl

Specifications

Absorbance 2000 (at lambda min.)
Beilstein 19, V,6, 461
Merck Index 15, 3076
Infrared Spectrum Authentic
Lambda Max 501 to 503nm (in 0.1N NaOH)
Formula Weight 401.2
Loss on Drying 5% max. (110°C)
Grade Extra Pure
Packaging Glass bottle
Density 0.79g/mL
Color Brown/Red to Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material 2', 7'-Dichlorofluorescein
Show More Show Less
compliance-icons
Product Identifier
  • 2'',7''-Dichlorofluorescein
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.