missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein 3',6'-diacetate, 97%

Product Code. 10419491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419491 5 g Glass bottle 5g
10419301 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419491 Supplier Thermo Scientific Acros Supplier No. 347250050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2044-85-1 | C24H14Cl2O7 | 485.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-85-1
Molecular Formula C24H14Cl2O7
Molecular Weight (g/mol) 485.27
MDL Number MFCD00037501
InChI Key VQVUBYASAICPFU-UHFFFAOYSA-N
PubChem CID 104913
IUPAC Name (6'-acetyloxy-2',7'-dichloro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(OC(C)=O)C=C1O2

Specifications

Melting Point 239°C to 246°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 19, IV, 2912
Infrared Spectrum Authentic
Formula Weight 485.27
Percent Purity ≥96.0%
Packaging Glass bottle
Color Colorless to White
Quantity 5 g
Physical Form Powder or Crystals
Chemical Name or Material 2', 7'-Dichlorofluorescein 3', 6'-diacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2',7'-Dichlorofluorescein 3',6'-diacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.