missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dichlorofluorenyl Bromomethyl Ketone, TRC

Product Code. 30382439
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382439 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382439 Supplier Toronto Research Chemicals Supplier No. TRCD43448025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,7-Dichlorofluorenyl Bromomethyl Ketone
CAS 53221-22-0
Molecular Formula C15H9BrCl2O
Quantity 25 mg
Synonym 2-Bromo-1-(2,7-dichloro-9H-fluoren-4-yl)ethanone, Bromomethyl 2,7-Dichloro-4-fluorenyl Ketone
SMILES Clc1ccc2c(Cc3cc(Cl)cc(C(=O)CBr)c23)c1
Molecular Weight (g/mol) 356.04
Formula Weight 353.9214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.