missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromonaphthalene, 99%

Product Code. 15412528
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412528 5 g 5g
15422528 250 mg 250mg
15402528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412528 Supplier Thermo Scientific Alfa Aesar Supplier No. H56097.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58556-75-5 | C10H6Br2 | 285.97 g/mol

It is an intermediate in dyestuffs and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in dyestuffs and pharmaceuticals.

Solubility
Soluble in acetonitrile.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58556-75-5
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00238580
InChI Key ODJZWBLNJKNOJK-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
PubChem CID 625355
IUPAC Name 2,7-dibromonaphthalene
SMILES BrC1=CC2=CC(Br)=CC=C2C=C1

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
UN Number UN3077
Beilstein 1862041
Solubility Information Soluble in acetonitrile.
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2,7-Dibromonaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.