missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9H-fluorene, TRC

Product Code. 30381404
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381404 10 g
30383420 25 g
30377793 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381404 Supplier Toronto Research Chemicals Supplier No. TRCD42618310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16433-88-8
Molecular Formula C13H8Br2
Molecular Weight (g/mol) 324.01
Synonym 2,7-Dibromofluorene
SMILES Brc1ccc2c(Cc3cc(Br)ccc23)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H8Br2/c14-10-1-3-12-8(6-10)5-9-7-11(15)2-4-13(9)12/h1-4,6-7H,5H2
Formula Weight 321.8993
Chemical Name or Material 2,7-Dibromo-9H-fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.