missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diaminofluorene dihydrochloride, 97%, Thermo Scientific™

Product Code. 10204261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10204261 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204261 Supplier Thermo Scientific Acros Supplier No. 338220010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13548-69-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 260°C to 310°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C13H12N2·2HCl
MDL Number MFCD00012535
Quantity 1 g
Synonym 9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: sparingly soluble in ethanol and acetone,insoluble in ether
InChI Key QJXYCUYLZDQRIN-UHFFFAOYSA-N
SMILES C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl
IUPAC Name 9H-fluorene-2,7-diamine;dihydrochloride
Molecular Weight (g/mol) 269.17
PubChem CID 11543576
Formula Weight 269.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 7-Diaminofluorene dihydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,7-Diaminofluorene dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.