missing translation for 'onlineSavingsMsg'
Learn More

2,7-Di-tert-butylfluorene, 98%, Thermo Scientific Chemicals

Catalog Number p-7036942
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
58775-05-6
C21H26
278.439
MFCD03093998
DFZYPLLGAQIQTD-UHFFFAOYSA-N
2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
4090082
2,7-ditert-butyl-9H-fluorene
CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C
This item is not returnable. View return policy
58775-05-6
C21H26
278.439
MFCD03093998
DFZYPLLGAQIQTD-UHFFFAOYSA-N
2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
4090082
2,7-ditert-butyl-9H-fluorene
CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C

It is used as an organic light emitting diode (OLED) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an organic light emitting diode (OLED) intermediate.

Solubility
Soluble in toluene, hot methanol.

Notes
Store at -20°C. Avoid heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 121°C to 124°C
Quantity 1 g
Solubility Information Soluble in toluene,hot methanol.
Formula Weight 278.43
Percent Purity 98%
Chemical Name or Material 2,7-Di-tert-butylfluorene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'