missing translation for 'onlineSavingsMsg'
Learn More

2,7-Di-tert-butylfluorene, 97+%

Product Code. 15421668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421668 5 g 5g
15411668 1 g 1g
15431668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421668 Supplier Thermo Scientific Alfa Aesar Supplier No. H55430.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58775-05-6 | C21H26 | 278.439 g/mol

It is used as an organic light emitting diode (OLED) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an organic light emitting diode (OLED) intermediate.

Solubility
Soluble in toluene, hot methanol.

Notes
Store at -20°C. Avoid heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58775-05-6
Molecular Formula C21H26
Molecular Weight (g/mol) 278.439
MDL Number MFCD03093998
InChI Key DFZYPLLGAQIQTD-UHFFFAOYSA-N
Synonym 2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
PubChem CID 4090082
IUPAC Name 2,7-ditert-butyl-9H-fluorene
SMILES CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C

Specifications

Melting Point 121°C to 124°C
Quantity 5 g
Solubility Information Soluble in toluene,hot methanol.
Formula Weight 278.43
Percent Purity 98%
Chemical Name or Material 2,7-Di-tert-butylfluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.