missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedimethanol, 98%

Product Code. 11377735
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11377735

Brand: Thermo Scientific Alfa Aesar L02527.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1195-59-1 | C7H9NO2 | 139.154 g/mol

2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-59-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00006351
InChI Key WWFMINHWJYHXHF-UHFFFAOYSA-N
Synonym 2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
PubChem CID 70957
IUPAC Name [6-(hydroxymethyl)pyridin-2-yl]methanol
SMILES C1=CC(=NC(=C1)CO)CO

Specifications

Melting Point 110°C to 114°C
Boiling Point 185°C (15 mmHg)
Quantity 5 g
Beilstein 116016
Solubility Information Soluble in water.
Formula Weight 139.15
Percent Purity 98%
Chemical Name or Material 2,6-Pyridinedimethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.