missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxy-1-benzofuran-3-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10130602
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10130602 10 g Amber Glass Bottle 10g
10213182 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10130602 Supplier Thermo Scientific Maybridge Supplier No. KM10844EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69716-05-8
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD02677734
InChI Key QCXJFLREQGIACT-UHFFFAOYSA-N
Synonym 2-6-methoxy-1-benzofuran-3-yl acetic acid, 2-6-methoxybenzofuran-3-yl acetic acid, 6-methoxy-1-benzofuran-3-yl acetic acid, 3-benzofuranacetic acid, 6-methoxy, 2-6-methoxybenzo b furan-3-yl acetic acid, maybridge3_004130, 4ddh, ms0, 6-methoxybenzofuran-3-acetic acid, 6-methoxybenzofuran-3-yl acetic acid
PubChem CID 736818
IUPAC Name 2-(6-methoxy-1-benzofuran-3-yl)acetic acid
SMILES COC1=CC2=C(C=C1)C(=CO2)CC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.2
Percent Purity 97%
Chemical Name or Material 2-(6-Methoxy-1-benzofuran-3-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.