missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine, 98+%

Product Code. 11488713
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
Unit Size:
100mL
2500mL
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488713 500 mL 500mL
11434050 25 mL 25mL
11478713 100 mL 100mL
11498713 2.5 L 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11488713 Supplier Thermo Scientific Alfa Aesar Supplier No. A10478.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-48-5 | C7H9N | 107.16 g/mol

2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-48-5
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006345
InChI Key OISVCGZHLKNMSJ-UHFFFAOYSA-N
PubChem CID 7937
ChEBI CHEBI:32548
IUPAC Name 2,6-dimethylpyridine
SMILES CC1=CC=CC(C)=N1

Specifications

Melting Point -7°C
Density 0.923
Boiling Point 143°C to 145°C
Flash Point 33°C (91°F)
Odor Unpleasant
Refractive Index 1.497
Quantity 500 mL
UN Number UN1993
Beilstein 105690
Sensitivity Hygroscopic
Merck Index 14,5616
Synonym 2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene
Solubility Information Miscible with water.
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material 2,6-Lutidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.