missing translation for 'onlineSavingsMsg'
Learn More

2,6-Divanillylidenecyclohexanone, TRC

Product Code. 30381994
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381994 1 mg 1mg
30382531 10 mg -
30382173 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381994 Supplier Toronto Research Chemicals Supplier No. TRCD4944781MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-23-7
Molecular Formula C22H22O5
Molecular Weight (g/mol) 366.4
Synonym 2,6-Bis[(4-hydroxy-3-methoxyphenyl)methylene]cyclohexanone, 2,6-Bis(4-hydroxy-3-methoxybenzylidene)cyclohexanone, Beveno, Curcumoid, Cyclovalon, Cyclovalone, Cycvalon, Cyqualon, DVC, Divanil, Divanilliden cyclohexanone, Divanon, Flavugal, GT 18, NSC 26727, NSC 652893, Tsikvalon, Vanilone
SMILES COc1cc(C=C2CCCC(=Cc3ccc(O)c(OC)c3)C2=O)ccc1O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C22H22O5/c1-26-20-12-14(6-8-18(20)23)10-16-4-3-5-17(22(16)25)11-15-7-9-19(24)21(13-15)27-2/h6-13,23-24H,3-5H2,1-2H3/b16-10+,17-11+
Chemical Name or Material 2,6-Divanillylidenecyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.