missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylisonicotinic acid, 97%, Thermo Scientific™

Catalog Number 10597624
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
38947-57-8
C18H13NO2
275.31
MFCD00418438
PWKUURSWFJBXGO-UHFFFAOYSA-N
2,6-diphenylisonicotinic acid, 2,6-diphenyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylicacid, 2,6-diphenyl, 4-pyridinecarboxylic acid, 2,6-diphenyl, 2 6-diphenylisonicotinic acid 97
3490694
2,6-diphenylpyridine-4-carboxylic acid
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
38947-57-8
C18H13NO2
275.31
MFCD00418438
PWKUURSWFJBXGO-UHFFFAOYSA-N
2,6-diphenylisonicotinic acid, 2,6-diphenyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylicacid, 2,6-diphenyl, 4-pyridinecarboxylic acid, 2,6-diphenyl, 2 6-diphenylisonicotinic acid 97
3490694
2,6-diphenylpyridine-4-carboxylic acid
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 283°C to 286°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 275.3
Percent Purity 97%
Chemical Name or Material 2, 6-Diphenylisonicotinic acid
compliance-icons
Product Identifier
  • 2,6-Diphenylisonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'