missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitroaniline, 98%, Thermo Scientific Chemicals

Catalog Number 11420063
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
606-22-4
C6H5N3O4
183.123
MFCD00007148
QFUSCYRJMXLNRB-UHFFFAOYSA-N
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
69070
2,6-dinitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]
This item is not returnable. View return policy
606-22-4
C6H5N3O4
183.123
MFCD00007148
QFUSCYRJMXLNRB-UHFFFAOYSA-N
benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
69070
2,6-dinitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]

2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY
Melting Point ∼136°C (decomposition)
Quantity 5 g
UN Number UN1596
Beilstein 2214886
Merck Index 14,3271
Solubility Information Insoluble in water.
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 2,6-Dinitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'