missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpyridin-3-ol, 97%, Thermo Scientific™

Product Code. 10190932
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10190932 1 g 1g
10361615 5 g 5g
10019341 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190932 Supplier Thermo Scientific Maybridge Supplier No. RH01494DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1122-43-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00228692
InChI Key AMQJAKKATFMFTR-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
PubChem CID 70732
IUPAC Name 2,6-dimethylpyridin-3-ol
SMILES CC1=CC=C(O)C(C)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethylpyridin-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.