missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpyrazine, 99%

Product Code. 11400568
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400568 5 g 5g
11410568 25 g 25g
11420568 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400568 Supplier Thermo Scientific Alfa Aesar Supplier No. B24687.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-50-9 | C6H8N2 | 108.144 g/mol

2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-50-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006148
InChI Key HJFZAYHYIWGLNL-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dimethyl, 3,5-dimethylpyrazine, 2,6-dimethylpiazine, 2,6-dimethylparadiazine, 2,6-dimethyl-1,4-diazine, ccris 2930, unii-n77q72c9i3, fema no. 3273, natural, 2,6-dimethyl pyrazine
PubChem CID 7938
IUPAC Name 2,6-dimethylpyrazine
SMILES CC1=CN=CC(=N1)C

Specifications

Melting Point 38°C to 43°C
Boiling Point 153°C to 154°C
Flash Point 52°C (125°F)
Odor Aromatic
Quantity 5 g
UN Number UN1325
Beilstein 1726
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylpyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.