missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpiperidine, predominantly cis, 99%

Product Code. 11454050
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11454050

Brand: Thermo Scientific Alfa Aesar B24524.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-03-0 | C7H15N | 113.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-03-0
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00066304
InChI Key SDGKUVSVPIIUCF-UHFFFAOYSA-N
Synonym nanofin, lupetidine, naniopinum, 2,6-lupetidine, lupetidin, nanophin, nanophine, nanophyn, piperidine, 2,6-dimethyl, naniopine
PubChem CID 68843
IUPAC Name 2,6-dimethylpiperidine
SMILES CC1CCCC(N1)C

Specifications

Melting Point <-20°C
Density 0.832
Boiling Point 126°C to 129°C
Flash Point 11°C (51°F)
Refractive Index 1.44
Quantity 5 g
UN Number UN2733
Beilstein 79827
Sensitivity Air Sensitive
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylpiperidine, predominantly cis
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.