missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpiperazine, 98%, Thermo Scientific™

Product Code. 15552283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15552283 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15552283 Supplier Thermo Scientific Acros Supplier No. 116671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 108-49-6
Melting Point 108.0°C to 113.0°C
Boiling Point 162.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C6H14N2
MDL Number MFCD07772435,MFCD00005956
Quantity 100 g
Beilstein 23,I,8
Synonym piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents
InChI Key IFNWESYYDINUHV-UHFFFAOYNA-N
SMILES CC1CNCC(C)N1
IUPAC Name 2,6-dimethylpiperazine
Molecular Weight (g/mol) 114.19
PubChem CID 66056
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylpiperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethylpiperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.