missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenylboronic acid, 98%

Product Code. p-2689864 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10179931

Brand: Thermo Scientific Acros 359270010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100379-00-8 | C8H11BO2 | 149.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100379-00-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01009693
InChI Key ZXDTWWZIHJEZOG-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
PubChem CID 583322
IUPAC Name (2,6-dimethylphenyl)boronic acid
SMILES CC1=CC=CC(C)=C1B(O)O

Specifications

Melting Point 105°C
CAS Min % 97.5
Color White
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 2, 6-Dimethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.