missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl Isothiocyanate, Mikromol™

Product Code. 30051638
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051638 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051638 Supplier Mikromol Supplier No. MM0515.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,6-Dimethylphenyl Isothiocyanate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dimethylphenyl Isothiocyanate
CAS 19241-16-8
Analyte or Component Names 2,6-Dimethylphenyl Isothiocyanate
Shipping Condition Room Temperature
Molecular Formula C9 H9 N S
Certifications/Compliance ISO 9001
Synonym 2,6-Dimethylphenyl Isothiocyanate, Xylazine Hydrochloride Imp. C (EP), Xylazine Imp. C (EP), 2-Isothiocyanato-1,3-dimethylbenzene, Isothiocyanic acid, 2,6-xylyl ester (6CI,8CI), 2,6-Dimethylphenyl isothiocyanate, 2,6-Dimethylphenyl thioisocyanate, 2,6-Xylyl isothiocyanate, NSC 172978, Xylazine Hydrochloride Impurity C, Xylazine Impurity C
InChI Formula InChI=1S/C9H9NS/c1-7-4-3-5-8(2)9(7)10-6-11/h3-5H,1-2H3
SMILES Cc1cccc(C)c1N=C=S
IUPAC Name 2-isothiocyanato-1,3-dimethylbenzene
Molecular Weight (g/mol) 163.24
Quantity 100 mg
Formula Weight 163.0456 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.