missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isocyanide

Product Code. 11337967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337967 1 g 1g
11347967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337967 Supplier Thermo Scientific Alfa Aesar Supplier No. 020748.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2769-71-3 | C9H9N | 131.18 g/mol

2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2769-71-3
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00013479
InChI Key DNJLFZHMJDSJFN-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide
PubChem CID 76009
IUPAC Name 2-isocyano-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1[N+]#[C-]

Specifications

Melting Point 73°C to 74°C
Quantity 1 g
Odor Unpleasant
Physical Form Crystalline
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 131.18
Chemical Name or Material 2,6-Dimethylphenyl isocyanide
compliance-icons
Product Identifier
  • 2,6-Dimethylphenyl isocyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Xylyl isocyanide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.