missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dimethylphenoxy)acetic Acid, TRC

Product Code. 30388156
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388156 Supplier Toronto Research Chemicals Supplier No. TRCD47647025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,6-Dimethylphenoxy)acetic Acid
CAS 13335-71-2
Molecular Formula C10 H12 O3
Quantity 25 g
Synonym Acetic acid, 2-(2,6-dimethylphenoxy)-, Acetic acid, (2,6-dimethylphenoxy)- (9CI), Acetic acid, (2,6-xylyloxy)- (6CI,8CI), 2-(2,6-Dimethylphenoxy)acetic acid, (2,6-Dimethylphenoxy)acetic acid, NSC 408599
InChI Formula InChI=1S/C10H12O3/c1-7-4-3-5-8(2)10(7)13-6-9(11)12/h3-5H,6H2,1-2H3,(H,11,12)
SMILES Cc1cccc(C)c1OCC(=O)O
IUPAC Name 2-(2,6-dimethylphenoxy)acetic acid
Molecular Weight (g/mol) 180.2
Formula Weight 180.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.