missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylmorpholine, 97%, mixture of isomers

Product Code. 11926811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11926811 100 g 100g
11916811 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926811 Supplier Thermo Scientific Acros Supplier No. 155341000
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 141-91-3 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-91-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key HNVIQLPOGUDBSU-UHFFFAOYSA-N
Synonym morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod
PubChem CID 110862
IUPAC Name 2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85.0°C
Density 0.9350g/mL
Boiling Point 147.0°C
Flash Point 48°C
Assay Percent Range 96% min. (mixture of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.447
Quantity 100 g
Specific Gravity 0.935
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethylmorpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethylmorpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.