missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylheptanoyl-CoA Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylheptanoyl-CoA Sodium Salt |
| Molecular Formula | C30 H52 N7 O17 P3 S . x(Na) |
| Quantity | 1 mg |
| Synonym | Coenzyme A S-(2,6-Dimethylheptanoate) Sodium Salt, 2,6-Dimethylheptanoyl Coenzyme A Sodium Salt |
| InChI Formula | InChI=1S/C30H52N7O17P3S.Na/c1-17(2)7-6-8-18(3)29(42)58-12-11-32-20(38)9-10-33-27(41)24(40)30(4,5)14-51-57(48,49)54-56(46,47)50-13-19-23(53-55(43,44)45)22(39)28(52-19)37-16-36-21-25(31)34-15-35-26(21)37;/h15-19,22-24,28,39-40H,6-14H2,1-5H3,(H,32,38)(H,33,41)(H,46,47)(H,48,49)(H2,31,34,35)(H2,43,44,45);/q;+1/t18?,19-,22-,23-,24-,28-;/m0./s1 |
| SMILES | [Na+].CC(C)CCCC(C)C(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@@H]1O[C@@H]([C@@H](O)[C@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23 |
| IUPAC Name | sodium;S-[2-[3-[[(2R)-4-[[[(2S,3R,4S,5S)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl]oxy-hydroxy-phosphoryl]oxy-2-hydroxy-3,3-dimethyl-butanoyl]amino]propanoylamino]ethyl] 2,6-dimethylheptanethioate |
| Molecular Weight (g/mol) | 930.7 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?