missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzoic acid, 99%

Product Code. 10296923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296923 25 g Glass bottle 25g
10464915 1 g Glass bottle 1g
10675412 5 g Glass bottle 5g
10563531 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10296923 Supplier Thermo Scientific Acros Supplier No. 169010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 632-46-2 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-46-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002483
InChI Key HCBHQDKBSKYGCK-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
PubChem CID 12439
ChEBI CHEBI:64827
IUPAC Name 2,6-dimethylbenzoic acid
SMILES CC1=C(C(=CC=C1)C)C(=O)O

Specifications

Melting Point 114.0°C to 117.0°C
CAS Min % 98.5
Color Cream to White
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Linear Formula (CH3)2C6H3COOH
Beilstein 01, 259
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 2, 6-Dimethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.