missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzeneboronic acid, 97%

Product Code. 11480468
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480468 1 g 1g
11490468 5 g 5g
11400478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480468 Supplier Thermo Scientific Alfa Aesar Supplier No. B24613.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 100379-00-8 | C8H11BO2 | 149.98 g/mol

It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100379-00-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01009693
InChI Key ZXDTWWZIHJEZOG-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
PubChem CID 583322
IUPAC Name (2,6-dimethylphenyl)boronic acid
SMILES CC1=CC=CC(C)=C1B(O)O

Specifications

Melting Point 120°C to 126°C (decomposition)
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.