missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylaniline, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Especificaciones
| Melting Point | 10°C to 12°C |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Density | 0.9800g/mL |
| Boiling Point | 216°C |
| Quantity | 5 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5591 to 1.5611 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- 2,6-Dimethylaniline
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 2,6-Xylidene
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido