Learn More
2,6-Dimethylaniline, 99%
Description
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.
Solubility
Slightly miscible with water. Miscible with ethanol and diethyl ether.
Notes
It is sensitive to air and light. Incompatible withacids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates and halogens.
Specifications
Specifications
| Melting Point | 11°C |
| Density | 0.981 |
| Quantity | 1000 g |
| Odor | Characteristic |
| Refractive Index | 1.56 |
| UN Number | UN1711 |
| Beilstein | 636332 |
| Merck Index | 14,10084 |
| Boiling Point | 216°C |
| Solubility Information | Slightly miscible with water. Miscible with ethanol and diethyl ether. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.