missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-p-benzoquinone, 99%

Product Code. 11493754
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493754 25 g 25g
11473754 1 g 1g
11483754 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493754 Supplier Thermo Scientific Alfa Aesar Supplier No. A11342.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 527-61-7 | C8H8O2 | 136.15 g/mol

2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-61-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001605
InChI Key SENUUPBBLQWHMF-UHFFFAOYSA-N
Synonym 2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone
PubChem CID 68241
IUPAC Name 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=C(C1=O)C

Specifications

Melting Point 69°C to 71°C
Quantity 25 g
Beilstein 2041345
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-p-benzoquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.